Learn More
Biomatik Corporation Rat Cytochrome C (CYCS) ELISA Kit
Supplier:  Biomatik Corporation EKU10777
Encompass_Preferred
Rat Cytochrome C (CYCS) ELISA Kit for detecting CYCS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62898. Gene ID: 100363502. Target Synonyms: Cyt-C; HCS; CYC; CytC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5629