Learn More
Biomatik Corporation Rat Cytochrome P450 11B1 (CYP11B1) ELISA Kit
Supplier:  Biomatik Corporation EKU03589
Encompass_Preferred
Rat Cytochrome P450 11B1 (CYP11B1) ELISA Kit for detecting CYP11B1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15393. Target Synonyms: CPN1; CYP11B; FHI; CYPXIB1; S11BH; 11-Beta-Hydroxylase; Cytochrome P450 Family 11 Subfamily B Polypeptide 1; Steroid 11-beta-hydroxylase; Cytochrome P-450c11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6830