Learn More
American Research Products Inc Rat Cytochrome P450 19A1 CYP1
Catalog No. 5000326872
Encompass
Rat Cytochrome P450 19A1 (CYP19A1) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26872 Supplier American Research Products Inc Supplier No. CSBEL006394RA
Rat Cytochrome P450 19A1 (CYP19A1) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 68 pg/mL Sample serum plasma tissue homogenates Method Sandwich