Learn More
American Research Products Inc Rat cytochrome P450 2E1 CYP2E
Catalog No. 5000328739
Encompass
Rat cytochrome P450 2E1 CYP2E1 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28739 Supplier American Research Products Inc Supplier No. CSBE09782R
Rat cytochrome P450 2E1 CYP2E1 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich