Learn More
Biomatik Corporation Rat Cytochrome P450 2E1 (CYP2E1) ELISA Kit
Supplier:  Biomatik Corporation EKU03606
Encompass_Preferred
Rat Cytochrome P450 2E1 (CYP2E1) ELISA Kit for detecting CYP2E1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05182. Gene ID: 25086. Target Synonyms: CPE1; CYPIIE1; P450-J; P450C2E; 4-nitrophenol 2-hydroxylase; Cytochrome P450 Family 2 Subfamily E 1; Cytochrome P450,Subfamily IIE (Ethanol-Inducible) Polypeptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5882