Learn More
American Research Products Inc Rat cytochrome P450 family 7
Catalog No. 5000328112
Encompass
Rat cytochrome P450 family 7 subfamily A polypeptide 1 (CYP7A1) ELISA kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 4 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28112 Supplier American Research Products Inc Supplier No. CSBEL006460RA
Rat cytochrome P450 family 7 subfamily A polypeptide 1 (CYP7A1) ELISA kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 4 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich