Learn More
Biomatik Corporation Rat Cytoglobin (CYGB) ELISA Kit
Supplier:  Biomatik Corporation EKU03615
Encompass_Preferred
Rat Cytoglobin (CYGB) ELISA Kit for detecting CYGB in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q921A4. Gene ID: 170520. Target Synonyms: HGB; STAP; Stellate Cell Activation-Associated Protein; Histoglobin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4614