Learn More
Biomatik Corporation Rat Cytokeratin 1 (CK1) ELISA Kit
Supplier:  Biomatik Corporation EKU05460
Encompass_Preferred
Rat Cytokeratin 1 (CK1) ELISA Kit for detecting CK1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6IMF3. Gene ID: 300250. Target Synonyms: KRT1; EHK1; K1; KRT1A; KRT1-A; Keratin 1; Epidermolytic Hyperkeratosis; Keratin, type II cytoskeletal 1; 67 kDa cytokeratin; Type-II keratin Kb1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8905