Learn More
Raybiotech Inc C-Series Rat Cytokine Antibody Array 1 Kit.
Catalog No. 501948778
Encompass_Preferred
Detects 19 Rat Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: CINC-2, CINC-3, CNTF, Fractalkine (CX3CL1), GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-4, IL-6, IL-10, LIX, Leptin, MCP-1 (CCL2), MIP-3 alpha (CCL20), beta-NGF, TIMP-1, TNF alpha, VEGF-A
Catalog No. 50-194-8778 Supplier Raybiotech Inc Supplier No. AARCYT12
Detects 19 Rat Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: CINC-2, CINC-3, CNTF, Fractalkine (CX3CL1), GM-CSF, IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-4, IL-6, IL-10, LIX, Leptin, MCP-1 (CCL2), MIP-3 alpha (CCL20), beta-NGF, TIMP-1, TNF alpha, VEGF-A