Learn More
Raybiotech Inc C-Series Rat Cytokine Antibody Array 2 Kit.
Catalog No. 501948781
Encompass_Preferred
Detects 34 Rat Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Activin A, Agrin, CD86 (B7-2), beta-NGF, CINC-1 (CXCL1), CINC-2, CINC-3, CNTF, Fas Ligand (TNFSF6), Fractalkine (CX3CL1), GM-CSF, ICAM-1 (CD54), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 R6 (IL-1 Rrp2), IL-2, IL-4, IL-6, IL-10, IL-13, Leptin, LIX, L-Selectin (CD62L), MCP-1 (CCL2), MIP-3 alpha (CCL20), MMP-8, PDGF-AA, Prolactin R, RAGE, TCK-1 (CXCL7), TIMP-1, TNF alpha, VEGF-A
Catalog No. 50-194-8781 Supplier Raybiotech Inc Supplier No. AARCYT22
Detects 34 Rat Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Activin A, Agrin, CD86 (B7-2), beta-NGF, CINC-1 (CXCL1), CINC-2, CINC-3, CNTF, Fas Ligand (TNFSF6), Fractalkine (CX3CL1), GM-CSF, ICAM-1 (CD54), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-1 R6 (IL-1 Rrp2), IL-2, IL-4, IL-6, IL-10, IL-13, Leptin, LIX, L-Selectin (CD62L), MCP-1 (CCL2), MIP-3 alpha (CCL20), MMP-8, PDGF-AA, Prolactin R, RAGE, TCK-1 (CXCL7), TIMP-1, TNF alpha, VEGF-A