Learn More
Raybiotech Inc G-Series Rat Cytokine Array 1 Kit.
Catalog No. 501950496
Encompass_Preferred
Semi-quantitative measurement of 10 rat cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: CINC-2, CINC-3, CNTF, Fractalkine (CX3CL1), GM-CSF, IL-1 alpha (IL-1 F1), IL-4, LIX, beta-NGF, VEGF-A
Catalog No. 50-195-0496 Supplier Raybiotech Inc Supplier No. GSRCYT11
Semi-quantitative measurement of 10 rat cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: CINC-2, CINC-3, CNTF, Fractalkine (CX3CL1), GM-CSF, IL-1 alpha (IL-1 F1), IL-4, LIX, beta-NGF, VEGF-A