Learn More
Raybiotech Inc G-Series Rat Cytokine Array 2 Kit.
Supplier:  Raybiotech Inc GSRCYT22
Encompass_Preferred
Semi-quantitative measurement of 10 rat cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: ICAM-1 (CD54), IFN-gamma, IL-1 beta (IL-1 F2), IL-10, IL-6, Leptin, L-Selectin (CD62L), MCP-1 (CCL2), TIMP-1, TNF alpha
Catalog No. 50-195-0499