Learn More
Raybiotech Inc G-Series Rat Cytokine Array 3 Kit.
Supplier:  Raybiotech Inc GSRCYT32
Encompass_Preferred
Semi-quantitative measurement of 27 rat cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: beta-NGF, CD86 (B7-2), CINC-1 (CXCL1), CINC-2, CINC-3, CNTF, Fractalkine (CX3CL1), GM-CSF, ICAM-1 (CD54), IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-10, IL-13, IL-2, IL-4, IL-6, LIX, L-Selectin (CD62L), MCP-1 (CCL2), PDGF-AA, Prolactin R, RAGE, TCK-1 (CXCL7), TIMP-1, TNF alpha, VEGF-A
Catalog No. 50-195-0501