Learn More
Raybiotech Inc G-Series Rat Cytokine Array 4 Kit.
Supplier:  Raybiotech Inc GSRCYT42
Encompass_Preferred
Semi-quantitative measurement of 40 rat cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: 4-1BB (TNFRSF9/CD137), Activin A, Adiponectin (ACRP30), CD80 (B7-1), CD48 (SLAMF2), CTACK (CCL27), Decorin, Eotaxin-1 (CCL11), EphA5, Erythropoietin, FGF-BP, Flt-3 Ligand, Galectin-1, Galectin-3, Gas 1, GFR alpha-1 (GDNF R), gp130, HGF, IL-1 R6 (IL-1 Rrp2), IL-1 ra (IL-1 F3), IL-2 R alpha, IL-3, IL-7, IL-17F, IL-22, JAM-A (CD321/F11R), MIP-1 alpha (CCL3), Neuropilin-1, Neuropilin-2, Nope, Notch-1, Notch-2, P-Cadherin, Prolactin, RANTES (CCL5), SCF, TIM-1 (KIM-1), TIMP-2, TREM-1, TWEAK R (TNFRSF12)
Catalog No. 50-195-0503