Learn More
Raybiotech Inc Quantibody Rat Cytokine Array 2 Kit.
Catalog No. 501950906
Encompass_Preferred
Detects 10 Rat Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: ICAM-1 (CD54), IFN-gamma, IL-1 beta (IL-1 F2), IL-10, IL-6, Leptin, L-Selectin (CD62L), MCP-1 (CCL2), TIMP-1, TNF alpha
Catalog No. 50-195-0906 Supplier Raybiotech Inc Supplier No. QARCYT22
Detects 10 Rat Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: ICAM-1 (CD54), IFN-gamma, IL-1 beta (IL-1 F2), IL-10, IL-6, Leptin, L-Selectin (CD62L), MCP-1 (CCL2), TIMP-1, TNF alpha