Learn More
Raybiotech Inc Quantibody Rat Cytokine Array 4 Kit.
Supplier:  Raybiotech Inc QARCYT42
Encompass_Preferred
Detects 40 Rat Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: 4-1BB (TNFRSF9/CD137), Activin A, Adiponectin (ACRP30), CD80 (B7-1), CD48 (SLAMF2), CTACK (CCL27), Decorin, Eotaxin-1 (CCL11), EphA5, Erythropoietin, FGF-BP, Flt-3 Ligand, Galectin-1, Galectin-3, Gas 1, GFR alpha-1 (GDNF R), gp130, HGF, IL-1 R6 (IL-1 Rrp2), IL-1 ra (IL-1 F3), IL-2 R alpha, IL-3, IL-7, IL-17F, IL-22, JAM-A (CD321/F11R), MIP-1 alpha (CCL3), Neuropilin-1, Neuropilin-2, Nope, Notch-1, Notch-2, P-Cadherin, Prolactin, RANTES (CCL5), SCF, TIM-1 (KIM-1), TIMP-2, TREM-1, TWEAK R (TNFRSF12)
Catalog No. 50-195-0910