Learn More
Raybiotech Inc Quantibody Rat Cytokine Array 67 Kit.
Catalog No. 501950901
Encompass_Preferred
A combination of 2 non-overlapping arrays (QAR-CYT-3 (27) & QAR-CYT-4 (40)) to quantitatively measure the concentration of 67 rat cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAR-CAA-67-1
Catalog No. 50-195-0901 Supplier Raybiotech Inc Supplier No. QARCAA671
A combination of 2 non-overlapping arrays (QAR-CYT-3 (27) & QAR-CYT-4 (40)) to quantitatively measure the concentration of 67 rat cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. View protein list on RayBiotech’s website – QAR-CAA-67-1