Learn More
Sino Biological Recombinant Rat Decorin Protein (His Tag)
Supplier:  Sino Biological 80656R08H200
Encompass_Preferred
A DNA sequence encoding the rat DCN (NP_077043.1 ) (Met1-Lys354) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2766