Learn More
Sino Biological Anti-DDR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80440T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat DDR1 (rR DDR1; Catalog#80440-R08H; Q6MG19; Met1-Thr413). DDR1 specific IgG was purified by Rat DDR1 affinity chromatography.
Catalog No. 50-241-0872