Learn More
Sino Biological Rat DDR1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80440R08H100
Encompass_Preferred
A DNA sequence encoding the rat DDR1 (Q6MG19) (Met1-Thr413) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2726