Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Decorin ELISA Kit

Catalog No. ERA8RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ERA8RBX5 5 x 96 Tests
ERA8RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ERA8RBX5 Supplier Invitrogen™ Supplier No. ERA8RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Rat Decorin quantitates rat Decorin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Decorin.

Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q01129
Assay Range 20-5000 pg/mL
Assay Sensitivity 20 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; Dermatan sulphate proteoglycans II (DSPG2); DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; Small leucine rich protein 1B; small leucine-rich protein 1B; unnamed protein productBone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; Dermatan sulphate proteoglycans II (DSPG2); DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; Small leucine rich protein 1B; small leucine-rich protein 1B; unnamed protein product;
Gene ID (Entrez) 29139
Gene Symbol DCN
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Decorin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.