Learn More
American Research Products Inc Rat Dehydroepiandrosterone DH
Catalog No. 5000329104
Encompass
Rat Dehydroepiandrosterone DHEA ELISA Kit from CUSABIO Detection Range 0 05 ng/mL-10 ng/mL Sensitivity 0 025 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29104 Supplier American Research Products Inc Supplier No. CSBE08227R
Rat Dehydroepiandrosterone DHEA ELISA Kit from CUSABIO Detection Range 0 05 ng/mL-10 ng/mL Sensitivity 0 025 ng/mL Sample serum plasma tissue homogenates Method Competitive