Learn More
American Research Products Inc Rat Dehydroepiandrosterone sul
Catalog No. 5000329406
Encompass
Rat Dehydroepiandrosterone sulfate DHEA-S ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29406 Supplier American Research Products Inc Supplier No. CSBE05106R
Rat Dehydroepiandrosterone sulfate DHEA-S ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma Method Sandwich