Learn More
Biomatik Corporation Rat Deiodinase, Iodothyronine, Type II (DIO2) ELISA Kit
Supplier:  Biomatik Corporation EKU03676
Encompass_Preferred
Rat Deiodinase, Iodothyronine, Type II (DIO2) ELISA Kit for detecting DIO2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70551. Gene ID: 65162. Target Synonyms: 5DII; D2; SelY; TXDI2; Thyroxine Deiodinase,Type II; Type-II 5'-deiodinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5593