Learn More
World Precision Instrument WPI Standard Rat Digital Dual Stereotaxic Instrument
SDP
Catalog No. 502539103
Encompass
10 Micron Resolution in All Three Directions
Building on WPI Rat Stereotaxic design, our Digital model offers greater speed of operation and increased accuracy. Fit with an easy-to-read digital display and sealed electronic sensors operation of the instrument is easy and precise. Simply move your probe to an established reference point (e.g. Bregma), zero the LED display and move to the desired coordinates.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
Catalog No. 50-253-9103 Supplier World Precision Instrument Supplier No. 505354
Add to Cart
Add to Cart
Request a Quote
10 Micron Resolution in All Three Directions
Building on WPI Rat Stereotaxic design, our Digital model offers greater speed of operation and increased accuracy. Fit with an easy-to-read digital display and sealed electronic sensors operation of the instrument is easy and precise. Simply move your probe to an established reference point (e.g. Bregma), zero the LED display and move to the desired coordinates.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.