Learn More
American Research Products Inc Rat dipeptidyl peptldase IV D
Catalog No. 5000330227
Encompass
Rat dipeptidyl peptldase IV DPPIV ELISA Kit from CUSABIO Detection Range 0 78 ng/mL - 50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30227 Supplier American Research Products Inc Supplier No. CSBE08519R
Rat dipeptidyl peptldase IV DPPIV ELISA Kit from CUSABIO Detection Range 0 78 ng/mL - 50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich