Learn More
Biomatik Corporation Rat Discs, Large Homolog 4 (DLG4) ELISA Kit
Supplier:  Biomatik Corporation EKU03762
Encompass_Preferred
Rat Discs, Large Homolog 4 (DLG4) ELISA Kit for detecting DLG4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31016. Gene ID: 29495. Target Synonyms: PSD95; SAP90; Postsynaptic density protein 95; Synapse-associated protein 90. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5261