Learn More
Sino Biological Anti-DLL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80013RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat DLL1 / Delta-like 1 (Catalog#80013-R08H; EDL99825.1; Met1-Ser534). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2399