Learn More
Lonza Walkersville Rat Neonatal Dorsal Root Ganglion (DRG) Neurons
Supplier:  Lonza Walkersville RDRG505
Encompass
Ready-to-use Clonetics Rat Dorsal Root Ganglion Neurons are suspensions of high quality sensory neurons prepared by standardized methods, and are ready for immediate culture.Each vial of dorsal root ganglia cells contains approximately 200,000 cells in 0.25 ml suspension. Cell death will occur during the first few days after plating and debris will be observed. Benefits:Ready-To-Use No tissue dissection or animal handling required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batchApplications:NeurogenesisNeurotransmitter FunctionGene ExpressionSignaling PathwaysElectrophysiologyNeurotoxicityDrug/Compound ScreeningAdvanced Cell Culture ModelsContent:≥200,000 cryopreserved cells per ampule
Catalog No. NC1986826