Learn More
Biomatik Corporation Rat Doublecortin Like Kinase 1 (DCLK1) ELISA Kit
Supplier:  Biomatik Corporation EKU12448
Encompass_Preferred
Rat Doublecortin Like Kinase 1 (DCLK1) ELISA Kit for detecting DCLK1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08875. Gene ID: 83825. Target Synonyms: DCDC3A; DCLK; DCAMKL1; Doublecortin And CaM Kinase-Like 1; Doublecortin domain-containing protein 3A; Doublecortin-like and CAM kinase-like 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7963