Learn More
Sino Biological Rat DSC2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 80266R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat DSC2 (rR DSC2; Catalog#80266-R08H; Q3T1K6; Met1-Pro694).
Catalog No. 50-237-3247