Learn More
Biomatik Corporation Rat Dual Oxidase 2 (DUOX2) ELISA Kit
Supplier:  Biomatik Corporation EKU03796
Encompass_Preferred
Rat Dual Oxidase 2 (DUOX2) ELISA Kit for detecting DUOX2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ES45. Gene ID: 79107. Target Synonyms: LNOX2; NOXEF2; THOX2; Thyroid oxidase 2; NADH/NADPH thyroid oxidase p138-tox; Dual Oxidase-Like Domains 2; Nicotinamide Adenine Dinucleotide Phosphate Oxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9003