Learn More
World Precision Instrument WPI Standard Rat Dual Stereotaxic Instrument, Non-Rupture Ear Bars
SDP
Catalog No. 502539145
Encompass
The Dual Arm WPI Rat Stereotaxic Instruments can decrease surgery times and help reduce potential time-consuming errors. A second manipulator arm is convenient, if not mandatory, in applications such as dual injections, bilateral cannulations, simultaneous stimulation and recording, etc.
NEW FEATURE: This is the ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9145 Supplier World Precision Instrument Supplier No. 505272
Add to Cart
Add to Cart
Request a Quote
The Dual Arm WPI Rat Stereotaxic Instruments can decrease surgery times and help reduce potential time-consuming errors. A second manipulator arm is convenient, if not mandatory, in applications such as dual injections, bilateral cannulations, simultaneous stimulation and recording, etc.
NEW FEATURE: This is the ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart