Learn More
World Precision Instrument WPI Standard Rat Stereotaxic Instrument
SDP
Catalog No. 502539196
Encompass
The Dual Arm WPI Rat Stereotaxic Instruments can decrease surgery times and help reduce potential time-consuming errors. A second manipulator arm is convenient, if not mandatory, in applications such as dual injections, bilateral cannulations, simultaneous stimulation and recording, etc.
NEW FEATURE: This is the ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9196 Supplier World Precision Instrument Supplier No. 505216
Add to Cart
Add to Cart
Request a Quote
The Dual Arm WPI Rat Stereotaxic Instruments can decrease surgery times and help reduce potential time-consuming errors. A second manipulator arm is convenient, if not mandatory, in applications such as dual injections, bilateral cannulations, simultaneous stimulation and recording, etc.
NEW FEATURE: This is the ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart