Learn More
BrainBits RAT E-18 CORTEX TISSUE CELLS
Catalog No. NC1238680
Encompass
E18 Rat Cortex. Microsurgically dissected Cortex tissue from E18 Sprague Dawley Rat. Fresh tissue and media are provided to iniate a culture of primary rat neurons. Guaranteed to Yield 2.0 Million Viable Cells.
Catalog No. NC1238680 Supplier BrainBits Supplier No. SDECX
E18 Rat Cortex. Microsurgically dissected Cortex tissue from E18 Sprague Dawley Rat. Fresh tissue and media are provided to iniate a culture of primary rat neurons. Guaranteed to Yield 2.0 Million Viable Cells.