Learn More
Raybiotech Inc Rat E-Selectin ELISA Kit
Supplier:  Raybiotech Inc ELRESELECTIN1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.13 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rat E-Selectin.
Catalog No. 50-195-0238