Learn More
BIOHIPPO Rat ECF Microsample
SDP
Catalog No. 5000313896
Encompass
Rat ECF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ECF in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ECF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ECF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ECF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13896 Supplier BIOHIPPO Supplier No. BHE15212009
Rat ECF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ECF in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ECF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ECF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ECF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color