Learn More
Biomatik Corporation Rat Echinoderm Microtubule Associated Protein Like Protein 2 (EML2) ELISA Kit
Supplier:  Biomatik Corporation EKU03828
Encompass_Preferred
Rat Echinoderm Microtubule Associated Protein Like Protein 2 (EML2) ELISA Kit for detecting EML2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P6T4. Gene ID: 192360. Target Synonyms: EMAP2; ELP70; Echinoderm MT-Associated Protein (EMAP)-Like Protein 70. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7728