Learn More
Sino Biological Anti-Ephrin-B1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80106RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Ephrin-B1 / EFNB1 (Catalog#80106-R08H; P52796; Met1-Thr229). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2828