Learn More
BIOHIPPO Rat eIF4E Microsample
SDP
Catalog No. 5000313719
Encompass
Rat eIF4E Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of eIF4E in serum plasma and other biological fluids It provides a detection range of 93 75-6000 pg/mL with a sensitivity of 26 86 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat eIF4E Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat eIF4E Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat eIF4E biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13719 Supplier BIOHIPPO Supplier No. BHE15210685
Rat eIF4E Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of eIF4E in serum plasma and other biological fluids It provides a detection range of 93 75-6000 pg/mL with a sensitivity of 26 86 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat eIF4E Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat eIF4E Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat eIF4E biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color