Learn More
BIOHIPPO Rat ELA2A Microsample
SDP
Catalog No. 5000315671
Encompass
Rat ELA2A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ELA2A in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 22 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ELA2A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ELA2A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ELA2A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15671 Supplier BIOHIPPO Supplier No. BHE15211034
Rat ELA2A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ELA2A in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 22 8 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ELA2A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ELA2A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ELA2A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color