Learn More
BIOHIPPO Rat ELA3B Microsample
SDP
Catalog No. 5000315672
Encompass
Rat ELA3B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ELA3B in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ELA3B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ELA3B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ELA3B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15672 Supplier BIOHIPPO Supplier No. BHE15211035
Rat ELA3B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ELA3B in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ELA3B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ELA3B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ELA3B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color