Learn More
Biomatik Corporation Rat Elastin Microfibril Interface Located Protein 1 (EMILIN1) ELISA Kit
Supplier:  Biomatik Corporation EKU10803
Encompass_Preferred
Rat Elastin Microfibril Interface Located Protein 1 (EMILIN1) ELISA Kit for detecting EMILIN1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3Z9E1. Target Synonyms: EMI; gp115; Elastin Microfibril Interfacer 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8286