Learn More
BIOHIPPO Rat ELN Microsample
SDP
Catalog No. 5000314059
Encompass
Rat ELN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ELN in serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ELN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ELN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ELN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14059 Supplier BIOHIPPO Supplier No. BHE15211220
Rat ELN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ELN in serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 9 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ELN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ELN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ELN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color