Learn More
Lonza Walkersville Rat Embryonic Dorsal Root Ganglion (eDRG) Neurons
Catalog No. NC1690958
Encompass
Ready to use Rat Embryonic Dorsal Root Ganglion (DRG) Neurons are suspensions of high quality sensory neurons prepared by standardized methods, and are ready for immediate culture.Benefits:Ready-To-Use No tissue dissection or animal handling required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batch Applications:NeurogenesisNeurotransmitter FunctionGene ExpressionSignaling PathwaysElectrophysiologyNeurotoxicityDrug/Compound ScreeningAdvanced Cell Culture ModelsContent:≥ 1,000,000 cryopreserved cells per ampule
This item is not returnable. View return policy
Catalog No. NC1690958 Supplier Lonza Walkersville Supplier No. REDRG515
Ready to use Rat Embryonic Dorsal Root Ganglion (DRG) Neurons are suspensions of high quality sensory neurons prepared by standardized methods, and are ready for immediate culture.Benefits:Ready-To-Use No tissue dissection or animal handling required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batch Applications:NeurogenesisNeurotransmitter FunctionGene ExpressionSignaling PathwaysElectrophysiologyNeurotoxicityDrug/Compound ScreeningAdvanced Cell Culture ModelsContent:≥ 1,000,000 cryopreserved cells per ampule