Learn More
Biomatik Corporation Rat Endoglin (ENG) ELISA Kit
Supplier:  Biomatik Corporation EKU03874
Encompass_Preferred
Rat Endoglin (ENG) ELISA Kit for detecting ENG in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.35ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6Q3E8. Gene ID: 497010. Target Synonyms: CD105; END; HHT1; ORW; ORW1; Osler-Rendu-Weber Syndrome 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7708