Learn More
Biomatik Corporation Rat Endostatin (ES) ELISA Kit
Supplier:  Biomatik Corporation EKU10102
Encompass_Preferred
Rat Endostatin (ES) ELISA Kit for detecting ES in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.23-100ng/ml. Sensitivity: 0.54ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9WUW5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8370