Learn More
Biomatik Corporation Rat Endothelial Cell Specific Molecule 1 (ESM1) ELISA Kit
Supplier:  Biomatik Corporation EKU11997
Encompass_Preferred
Rat Endothelial Cell Specific Molecule 1 (ESM1) ELISA Kit for detecting ESM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97682. Gene ID: 64536. Target Synonyms: Endocan. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6197