Learn More
Cell Biologics Complete Endothelial Cell Medium /w Kit – 500 ML
SDP
Catalog No. 501048384
Encompass_Preferred
(500 ml Basal medium with the growth factor supplement) Endothelial Cell Medium Supplement Kit, Cat No M1266-Kit includes: 0.5 ML EGF, 0.5 ML VEGF, 5.0 ML Antibiotic-Antimycotic Solution, 10.0 ML FBS
Catalog No. 50-104-8384 Supplier Cell Biologics Supplier No. M1266
(500 ml Basal medium with the growth factor supplement) Endothelial Cell Medium Supplement Kit, Cat No M1266-Kit includes: 0.5 ML EGF, 0.5 ML VEGF, 5.0 ML Antibiotic-Antimycotic Solution, 10.0 ML FBS