Learn More
American Research Products Inc Rat Endothelial nitric oXide s
Catalog No. 5000330210
Encompass
Rat Endothelial nitric oXide synthase eNOS/NOS3 ELISA Kit from CUSABIO Detection Range 15 6 IU/mL-1000 IU/mL Sensitivity 3 9 IU/mL Sample serum plasma cell culture supernates cell lysates Method Sandwich
Catalog No. 50-003-30210 Supplier American Research Products Inc Supplier No. CSBE08323R
Rat Endothelial nitric oXide synthase eNOS/NOS3 ELISA Kit from CUSABIO Detection Range 15 6 IU/mL-1000 IU/mL Sensitivity 3 9 IU/mL Sample serum plasma cell culture supernates cell lysates Method Sandwich